Would you like to log in again?
ToxOut™ Rapid Endotoxin Removal Kit, BioVision
Supplier: BioVision
AB239707-5KIT
75837-834EA 468.92 USD InStock
75837-834
ToxOut™ Rapid Endotoxin Removal Kit, BioVision
Protein Extraction Protein Extraction Kits
Quickly and effectively eliminate endotoxins to < 0.05 EU/ml in solutions containing proteins or pharmacologically important components.
  • High Binding Capacity: up to 9 x 108 EU/ml resin
  • High Sample Recovery: ~90% recovery with protein solution samples
  • Each column can be reused up to 5 times
  • Rapid removal: only require 5 minutes for each removal cycle
Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. A single E.coli bacterium contains ~2 million LPS molecules (2 – 20 fg/cell). During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. In vitro, endotoxin causes a variety of problems in cell-based research. In vivo, endotoxin may cause various side effects, including inflammatory response, organ failure or septic shock in host organisms. Therefore, it is critical to remove endotoxin from samples and products. BioVision‘s ToxOutTM Rapid Endotoxin Removal Kit can quickly and effectively eliminate endotoxins to < 0.05 EU/ml in solutions containing proteins or pharmacologically important components via the immobilized polymyxin B, which is known for capturing endotoxin and preventing toxic effects.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty