Would you like to log in again?
StemBoost™, Neuronal Cell Induction Cocktail Set (100X), Sterile-Filtered, BioVision
Supplier: BioVision
AB286239-1ML AB286239-5X1ML
75837-944EA 436.66 USD InStock
75837-944 75837-946
StemBoost™, Neuronal Cell Induction Cocktail Set (100X), Sterile-Filtered, BioVision
Cells
StemBoost™ Neuronal Cell Induction Cocktail Set (100X, in DMSO), Sterile-Filtered is a convenient, sterile-filtered cocktail solution.
  • Convenient
  • Sterile-filtered cocktail solution
Contains the following six small molecules: CHIR99021, a GSK-3β inhibitor, 0.3 mM; RepSox, a TGFβ inhibitor, 0.1 mM; Forskolin, a cAMP activator, 1.0 mM; SP600125, a JNK inhibitor, 1.0 mM; Gö6983, a PKC inhibitor, 0.5 mM and Y-27632 HCl , a ROCK inhibitor. Also provided separately is a sterile-filtered solution (100X, in DI water) of the HDAC inhibitor Valproic Acid Sodium Salt (50 MM).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty