Would you like to log in again?
Human Recombinant MMP-9 (from E. coli)
Supplier: BioVision
AB285982-1000UNITS AB285982-500UNITS
75838-000EA 766.09 USD InStock
75838-000 75837-998
Human Recombinant MMP-9 (from E. coli)
Enzymes
Matrix metallopeptidase 9 (MMP-9), also known as 92 kDa type IV collagenase, 92 kDa gelatinase or gelatinase B, is the mostly studied MMP, due to its fundamental role in cancer biology, autoimmune disease, and other conditions. This enzyme degrades various substrates including gelatin, collagen types IV and V, and elastin. MMP-9 is structurally a multi-domain metalloenzyme, composed of a prodomain, a catalytic domain, a gelatin binding domain, a metal-binding domain, and a carboxyl terminal hemopexin like domain.
  • Gene ID: 4319
  • Accession number: P14780
This active human MMP-9 is composed of the catalytic domain, a gelatin binding domain, and a metal binding domain (AA 107-457). The protein was expressed in E.coli and purified and refolded using proprietary techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty