Would you like to log in again?
VWR TIP PIPETTOR PRO STYLE STEP 0.2ML
Supplier: VWR International
VWR* Pro style step Pipettor Tip 0.2mL^ VWR* Pro style step Pipettor Tip, proper for viscous, infectious and volatile liquids, Reliable and repeatable results through automated, individual tests of every single tip, for steppers, Size: 0.2mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR TIP PIPETTOR PRO STYLE STEP 0.2ML 75838-196 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty