Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® High Speed Microcentrifuge

Supplier: VWR
VWR®
C1603-VWR
75838-336EA 2033.2 USD InStock
75838-336
VWR® High Speed Microcentrifuge
Centrifuges Microcentrifuges
The compact, affordable VWR® High Speed Microcentrifuge is capable of high speed performance up to 15500 rpm/16100×g.

  • Compact, less than 9" Ø
  • Near-silent operation
  • Large LCD with rcf conversion
  • Decelerated in just 10 seconds
  • Capacity: 12×1.5/2.0 ml

A solid, high-grade aluminum rotor is including and accepts microtubes (1.5 to 2.0 ml).

Time and speed settings are easily adjustable on a large LCD display and the speed can be set or viewed in rpm or rcf (relative centrifugal force). A safety imbalance mechanism is designed to stop the centrifuge if started out of balance and the locking lid cannot be opened unless the rotor has come to a complete stop. Other features include fast acceleration and deceleration (11 s), and reduced temperature buildup within the samples, even for extended runs at the maximum speed.

The rotor also includes a convenient snap-on plastic lid to reduce the risk of contamination.

2 year warranty.

Certifications: CE marked.

Delivery information: Supplied with rotor.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty