Would you like to log in again?
Avantor SterilEnz®-II/AT: Pre-Gasketed Sanitary Fittings for Single-Use Systems, Avantor Fluid Handling
Supplier: Avantor Fluid Handling
2AT-816-TC-10 2AT-616-TC-10 2AT-1216-TC-10 2AT-608-TC-10 2AT-608-TC-10 2AT-408-TC-10 2AT-812-TC-10 2AT-416-TC-10
75838-680CS 1384.65 USD InStock
75838-680 75838-681 75838-670 75838-672 75838-674 75838-676 75838-678 75838-668
Avantor SterilEnz®-II/AT: Pre-Gasketed Sanitary Fittings for Single-Use Systems, Avantor Fluid Handling
Fittings and Connectors
SterilEnz®-II/AT connectors come with a platinum cured, medical grade silicone gasket mechanically attached to the fitting face.
  • Standard sanitary clamp fitting, per ASME BPE Specifications
  • Class VI Compliant materials; certificate included
  • Won't leak or blow-off like stepped barbs can
  • Mono-Barb design
  • Zero mold parting lines on all critical sealing surfaces
  • Superior tooling
  • Gamma-stable, 25 - 40 kGY
  • For use in irradiated disposable systems
  • Autoclaveable at 123 °C for 30 minutes
  • Certified free from any animal derived components or processes
  • Animal-free
SterilEnz®-II/AT fittings are made rugged inert polypropylene suitable for either autoclaving or gamma-irradiation. The new II/AT series is available in seven fitting sizes.
Certifications: USP <88>: Class VI Biological reactivity tests, in vivo
USP <87>: Cytotoxicity, MEM Elution, in vitro
USP <661>: Physicochemical Test for plastics
USP <1207>: Sterile Product Packaging, hose barb integrity evaluation, via microbial immersion
ASTM F756-00 : Rabbit blood cell hemolysis
SterilEnz®-II/AT are certified ADCF (animal derived component free).
All SterilEnz®-II/AT products are assembled inside a certified ISO 7 (Class 10000) environment.
Protected by U.S. Patent 7,950,700, other patents pending. SterilEnz® is a registered trademark of PAW BioScience Products, LLC.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty