Would you like to log in again?
STERILENZ 3/8INHB X MINI TC PP W/ GASKET
Supplier: Avantor Fluid Handling
SterilEnz*-II/AT, 3/8in HB x Mini Flange Adapter^ SterilEnz*-II/AT, 3/8in HB x Mini Flange Adapter, Standard Sanitary Fittings for Single-Use Systems, Material: polypropylene, with Silicone gasket, Class VI Compliant Materials, Mono-Barb Design, Superior Tooling, Gamma-Stable, Autoclaveable
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STERILENZ 3/8INHB X MINI TC PP W/ GASKET 75838-680 2AT-608-TC-10 Case of 100 Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty