Would you like to log in again?
Globe Scientific Glue Seal Specimen Bags, Globe Scientific Inc
Supplier: Globe Scientific
4921
75838-694CS 289.59 USD InStock
75838-694
Globe Scientific Glue Seal Specimen Bags, Globe Scientific Inc
Bags Sample Bags
The special glue closure of our specimen transport bags offer a liquid-tight tamper evident seal.
  • 2 mil gauge polyethylene (PE)
  • Attached document compartment
  • Glue closure tamper evident seal
  • Imprinted with biohazard symbol
  • Perfect for the transport of drug testing products
Simply pull off the liner to expose the glue adhesive and press to close.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty