Would you like to log in again?
Neurotrophin 3 (NT3) Rapid ELISA Kit: Human, Rat and Mouse, Biosensis®
Supplier: Biosensis
BEK-2221-2P BEK-2221-1P
75838-790KT 826 USD InStock
75838-790 75838-788
Neurotrophin 3 (NT3) Rapid ELISA Kit: Human, Rat and Mouse, Biosensis®
Assays ELISAs
The Biosensis NT3 RapidTM enzyme-linked immune-sorbent assay (ELISA) Kit is a sandwich ELISA that allows the specific, fast and reliable quantification of NT3 in less than 4 hours in cell culture supernatants and human plasma (EDTA and citrate) only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application, in particular human plasma.
Accurate quantification of NT3 in human plasma requires a heterophilic antibody (HA) blocker which can be purchased separately (BL-004-500). This ELISA kit consists of a pre-coated polyclonal anti-NT3 capture antibody, a biotinylated monoclonal anti-NT3 detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin.
The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product, which is directly proportional to the concentration of NT3 present in samples and protein standards. A human NT3 positive control (QC sample) is provided to assure consistent assay performance. This NT3 RapidTM ELISA kit employs a recombinant human NT3 standard. NT3 is highly conserved and nearly identical in many species.
The assay's capture and detection antibodies detect NT3 from other species including mouse, rat and monkey, thus it is expected that this NT3 Rapid™ ELISA will also react with those species and possibly more. The antibodies used in this kit bind to epitopes within the mature domain of NT3. Thus, this ELISA kit may detect the full-length pro-form of NT3. This kit has not been tested for other applications. Sufficient amount of NT3 standard is supplied to allow for spike- and recovery experiments in order to validate this ELISA assay for other sample matrices if required.
Delivery information: The ELISA kit box contains 1 or 2×96-well pre-coated strip plate, protein standards, QC sample, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: For research use only. Not to be used in diagnostic or clinical procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty