Would you like to log in again?
Multi-Neurotrophin Rapid Screening ELISA Kit, Mouse, Biosensis®
Supplier: Biosensis
BEK-2231-SET
75838-798KT 643 USD InStock
75838-798
Multi-Neurotrophin Rapid Screening ELISA Kit, Mouse, Biosensis®
Assays ELISAs
The biosensis® Multi-Neurotrophin RapidTM Screening ELISA kit has been designed to allow rapid screening and quantification of mouse NGF, BDNF, NT3 and NT4/5 in cell culture supernatants, lysates and brain extracts only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application.
This two-plate kit consists of four sets of 6 strips for each Neurotrophin, allowing for the assay of 16 samples per Neurotrophin tested and a full range of standards, all run in duplicate. It provides the identical sensitivities and ranges that are achieved in the complete, individual ELISA kits, thus allowing easy progression into the larger individual assay sizes when needed.
The Multi-Neurotrophin RapidTM Screening ELISA kit therefore presents a cost-effective way to quickly screen multiple samples, which can then be published or used prior to a more extensive analysis with the individual kits. The kit comes complete with all detection reagents and is ready to use. Individual coated strips are provided, and each set of standards and detection antibodies are color-coded.
Delivery information: The ELISA kit box contains 2×96-well pre-coated strip plates (6 strips/48 wells per neurotrophin target), protein standards, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: For research use only. Not for diagnostic and clinical purposes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty