Would you like to log in again?
Mature BDNF Rapid ELISA Kit, Biosensis®
Supplier: Biosensis
BEK-2211-2P BEK-2211-1P
75838-764KT 826 USD InStock
75838-764 75838-838
Mature BDNF Rapid ELISA Kit, Biosensis®
Assays ELISAs
The Mature BDNF RapidTM ELISA Kit is a sandwich ELISA that allows the quantification of mature BDNF in less than 3 hours in cell culture supernatants, serum, plasma (citrate and EDTA), pig serum, cell lysates, brain extracts, human milk and sheep CSF only if used as directed, with a simplified protocol and no loss of sensitivity or specificity.
Please refer to the kit protocol for specific use instructions for each substrate application, in particular blood samples, human milk and CSF. Note that accurate quantification of BDNF in human milk requires a secretory IgA (sIgA) blocker which can be purchased separately. This ELISA kit has been tested in independent research laboratories and found to achieve highest reproducibility with intra- and inter-assay CVs as low as 1% and 5%, respectively.This ELISA kit consists of a pre-coated mouse monoclonal anti-mature BDNF capture antibody, a biotinylated anti-mature BDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin.
The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a coloured reaction product which is directly proportional to the concentration of mature BDNF present in samples and protein standards. This Mature BDNF ELISA kit employs a recombinant human mature BDNF standard approved by the World Health Organization.
The amino acid sequence of mature BDNF is identical for human, mouse, rat and a number of other species. This kit therefore is suitable to measure mature BDNF in all these species and uses the same antibodies and antigen.This ELISA kit has not been tested for other applications.
Delivery information: The ELISA kit box contains 1 or 2×96-well pre-coated strip plates, protein standards, QC sample, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: It has been configured for research use only and is not to be used for diagnostic or clinical procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty