Would you like to log in again?
GDNF Rapid ELISA Kit: Human, Biosensis®
Supplier: Biosensis
BEK-2222-1P BEK-2222-2P
75838-792KT 429 USD InStock
75838-792 75838-846
GDNF Rapid ELISA Kit: Human, Biosensis®
Assays ELISAs
The Biosensis GDNF RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the quantification of GDNF in less than 4 hours in cell culture supernatants, cell lysates, and guinea pig serum only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application.
This ELISA kit consists of a pre-coated anti-GDNF capture antibody, a biotinylated anti-GDNF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’- tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of GDNF present in samples and protein standards.
This GDNF ELISA kit employs a recombinant human GDNF standard obtained from the National Institute for Biological Standards and Control (NIBSC) and is therefore designed to accurately measure the human form of GDNF. Note that the antibodies used in this kit cross-react with guinea pig, rat and mouse GDNF. Guinea pig GDNF has been quantified in serum using the Human GDNF RapidTM ELISA kit, since guinea pig GDNF protein shares closest amino acid sequence homology with human GDNF. Protein levels can be expressed as “human GDNF equivalents”.
This kit has not been tested for other applications.
Delivery information: The ELISA kit box contains 1 or 2 x 96-well pre-coated strip plate, protein standards, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: It has been configured for research use only and is not to be used in diagnostic or clinical procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty