Would you like to log in again?
proNGF Rapid ELISA Kit: Human, Biosensis®
Supplier: Biosensis
BEK-2226-2P BEK-2226-1P
75838-850KT 826 USD InStock
75838-850 75838-848
proNGF Rapid ELISA Kit: Human, Biosensis®
Assays ELISAs
The Biosensis proNGF RapidTM enzyme-linked immunosorbent assay (ELISA) Kit is a sandwich ELISA that allows the quantification of full-length proNGF protein in less than 4 hours in human serum, heparin-plasma, cell supernatants and lysates only if used as directed. Please refer to the kit protocol for specific use instructions for each substrate application.
This ELISA kit consists of a pre-coated anti-proNGF capture antibody, a biotinylated anti-proNGF detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a colored reaction product which is directly proportional to the concentration of proNGF present in samples and protein standards. A human proNGF positive control (QC sample) is provided to assure consistent assay performance.
This proNGF ELISA kit contains a recombinant human proNGF standard expressed in E. coli. This ELISA kit does not cross-react with the mouse form of proNGF, and due to sequence homology of mouse and rat proNGF is not expected to detect rat proNGF. The antibodies used in this ELISA kit bind epitopes within the pro-domain of the protein and therefore recognize proNGF and the pro-domain peptide, but do not cross-react with mature NGF!
Note that accurate proNGF quantification in human serum requires the addition of Heterophilic Antibody Blocker BL-003-1000 provided in the kit, and available for purchase separately. This kit has not been tested for other applications. Sufficient amount of proNGF standard is supplied to allow for spike- and recovery experiments in order to validate this ELISA assay for other sample matrices if required.
Delivery information: The ELISA kit box contains 1 or 2 x 96-well pre-coated strip plate, protein standards, QC sample, detection reagents, heterophilic antibody blocker, wash and sample buffers, substrate buffer and detailed protocols.
Caution: This kit has been configured for research use only and is not to be used in diagnostic or clinical procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty