Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Kanamycin, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z7191
75838-868EA 31.94 USD InStock
75838-868
Kanamycin, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
Differentiates anaerobic bacteria on the basis of susceptibility to kanamycin.
  • Each cartridge contains 50 disks
  • Spring-loaded for smooth and reliable dispensing
  • Differentiates anaerobic bacteria
Differentiates anaerobic bacteria on the basis of susceptibility to kanamycin. Each cartridge contains 50 disks that are spring-loaded for smooth and reliable dispensing from a single metal disk dispenser.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty