Would you like to log in again?
TL23 Series Benchtop Turbidimeters, Hach®
Supplier: Hach
Hach®
LPV444.99.00310 LPV444.99.00310 LPV444.99.00320 LPV4449100210 LPV444.99.00120 LPV444.99.00210
75838-960EA 9425.5 USD InStock
75838-960 75838-962 CA75838-962 75838-944 CA75838-944 75838-946
TL23 Series Benchtop Turbidimeters, Hach®
Turbidity Meters
The TL23 series benchtop turbidimeters for a wide range of turbidity applications feature the trusted technology of Hach’s legacy benchtop turbidimeters and improved functionality to simplify testing.
  • Trusted measurement for high range turbidity applications; simplified
  • Improved and intuitive design
  • A smart device for more reliable measurements
  • Easy to use, easy to be right
These features include a color touch screen, a USB port for data transfer, self-diagnostics for quality assurance, and single point calibration for measurements under 40 NTU/FNU.
The TL23 series benchtop turbidimeters are available in four different models: two models with tungsten lamp at 460 nm, and two models with LED light source at 860 nm.
Trusted measurement for high range turbidity applications; simplified–The new TL23 series laboratory turbidimeters blend trusted technology and improved features to simplify testing in the most demanding industrial and wastewater applications.
Improved and intuitive design–the TL23 series’ large full color touch screen display and intuitive user interface accelerate setup, calibration and measurement. An easy interface and guided procedures give you confidence in your results.
The TL23 series provides everything you need at your fingertips. With a USB port for easy data export, sample identification for traceability, and self-diagnostics for quality assurance, Hach® makes it easy to be right.
Ordering information: Includes TL23 turbidimeter, silicone oil, oiling cloth, USEPA filter assembly, 1" sample cells (30 ml) with caps (6×), gelex secondary turbidity standardization kit, stablcal calibration kit, power supply, power cord, dust cover.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty