Would you like to log in again?
VIAL AMB BORO SC 8-425MM 2ML PK100
Supplier: MicroSolv Technology Corp
BASIK* Snap/Crimp top, 2ml,12x32mm, Glass, 1,000 series, Amber, plain^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL AMB BORO SC 8-425MM 2ML PK100 75839-412 9582S-0DAV Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty