Would you like to log in again?
SMART Digest™ ImmunoAffinity (IA) Protein G Kits
Supplier: Thermo Scientific
SMART Digest™
60112-104 60112-103 60112-101 60112-102
75839-558EA 2000.95 USD InStock
75839-558 75839-556 75839-562 75839-560
SMART Digest™ ImmunoAffinity (IA) Protein G Kits
Protein Purification Protein Purification Kits
Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well using Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Kits.
  • For peptide quantitation
  • Solving the challenges of low-level quantitation in complex matrices
  • Immunocapture and protein digestion in a single well
  • Unique design reduces workflow to hours
  • Capture: Protein G
  • Cleavage: C-terminal side of lysine and arginine amino acids
  • Enzyme: Trypsin
These kits were designed to remove the challenges often associated the process of biomarker quantitation by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight. The SMART Digest IA Protein G Kits provide targeted purification of immunoglobulins by utilizing the strong affinity for binding to the Fab and Fc portions of the molecule.
The need to identify low-level biomarkers is forcing the analytical community to deal with extremely high levels of analyte and sample complexity, which makes quantitation challenging. As these proteins are often present at low levels in complex biological matrices, an immunoaffinity capture step is typically employed to purify the sample and increase sensitivity. This IA step is often followed by protein digestion. Current processes are often time-consuming, multistep, and prone to irreproducibility. SMART Digest IA kits solve these problems.
In SMART Digest IA Kits, the immunocapture reagents and the temperature-activated, thermally stable trypsin are co-immobilized onto a resin. Following enrichment, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions, resulting in a workflow which takes hours rather than overnight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty