Would you like to log in again?
SMART Digest™ ImmunoAffinity (IA) Streptavidin Kits
Supplier: Thermo Scientific
SMART Digest™
60110-101 60110-102 60110-104 60110-103
75839-578EA 2261.18 USD InStock
75839-578 75839-432 75839-576 75839-580
SMART Digest™ ImmunoAffinity (IA) Streptavidin Kits
Protein Purification Protein Purification Kits
Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well using Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Streptavidin Kits.
  • The kit purifies various target biomolecules via the formation of strong streptavidin-biotin bond
  • Capture: Streptavidin
  • Cleavage: C-terminal side of lysine and arginine amino acids
  • Enzyme: Trypsin
  • For use with (Application): Peptide quantitation
These kits were designed to remove the challenges often associated with the process of biomarker quantitation by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight.
The need to improve the ability to identify low-level biomarkers is forcing the analytical community to deal with extremely high levels of analyte and sample complexity, which makes quantitation challenging. As these proteins are often present at low levels in complex biological matrices, an immunoaffinity capture step is typically employed to purify the sample and increase sensitivity. This step is often followed by protein digestion. Current processes are often time consuming, multistep, and prone to irreproducibility.
In SMART Digest IA Kits, the immunocapture reagents and the temperature-activated, thermally stable trypsin are co-immobilized onto a resin. Following enrichment, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions, resulting in a workflow which takes hours rather than overnight.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty