Would you like to log in again?
VWR® Maximum Registering/Autoclave Bi-Metal Thermometers
Supplier: VWR
61315-0011 60215-0011
75839-584CS 369.72 USD InStock
75839-584 75839-570
VWR® Maximum Registering/Autoclave Bi-Metal Thermometers
Thermometers Analog Thermometers
This durable bi-metal thermometer has a maximum registering pointer (red) that is moved up by the temperature measuring pointer (black) to indicate the highest temperature achieved, and remains there until manually reset.
  • Threaded connection (1/4” NPT) screws directly into in-line processes; can be used to mount thermometer on included stand
  • Waterproof and pressurized; withstands autoclave cycles and has a removable protective silicone sleeve to prevent moisture from making contact with the dial
  • Constructed from durable 304 stainless steel with a glass lens; withstands pressure, high temperatures and rust
  • Probe length: 50mm (2")
  • Tested and inspected by our USA quality team
  • Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and instructions
Also available with an individual Thermometer Calibration Report.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty