These useful silica-hydride HPLC columns have an amide functional group bonded directly to the particle with a direct silicon-carbon bond.
- Excellent for sugars and other carbohydrates
- Use in ANP or RP
The Cogent Amide phase is very stable and is recommended for Aqeous Normal Phase(ANP) or Reversed-Phase (RP) separations of biomolecules or macromolecules including carbohydrates, peptides, polysaccharides or tryptic digests.