Would you like to log in again?
COLUMN COGENT UDC-CHOL 4UM 75MMX21.2MM
Supplier: MicroSolv Technology Corp
Column Cogent Udc-Cholesterol Hplc Pore:100 armstrong, Particle size: 4um, Dimension: 75MMX21.2MM^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN COGENT UDC-CHOL 4UM 75MMX21.2MM 75839-760 69069-P21-75 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty