Would you like to log in again?
COLUMN COGENT DIOL 2.O 2.2UM 150MMX1.0MM
Supplier: MicroSolv Technology Corp
Column Cogent Diol Hplc Pore Size 120 armstrong, Particle size: 2.O 2.2um, Dimension: 150MMX1.0MM^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN COGENT DIOL 2.O 2.2UM 150MMX1.0MM 75839-920 40260-15P-1 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty