Would you like to log in again?
COLUMN CGNT PHEYNYL HYD 4UM 3.0MMX20MM
Supplier: MicroSolv Technology Corp
Column Cogent Pheynyl Hydride hplc, Pore:100 armstrong, Particle size: 4um, Dimension: 3.0MMX20MM^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN CGNT PHEYNYL HYD 4UM 3.0MMX20MM 75839-932 69020-02P-3 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty