Would you like to log in again?
KIT STROBOSCOPE LED NOVA-PRO 100
Supplier: MONARCH INSTRUMENT
Nova-Pro 100 Kit^ Nova-Pro 100 Kit, Nova-Pro 100 Stroboscope, Includes: standard Li-ion battery pack, battery charging station with interchangeable wall plugs, operation manual and plastic latching carry case, Display: 6 Digit Numeric /5 Digit Alphanumeric LCD, Tachometer Mode: 0-999,999 RPM
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT STROBOSCOPE LED NOVA-PRO 100 75840-608 6241-011 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty