Would you like to log in again?
KIT STROBOSCOPE LED 115/230VAC EURO PLUG
Supplier: MONARCH INSTRUMENT
Nova-Pro 100 AC Kit^ Nova-Pro 100 AC LED Stroboscope Kit, Includes: 115/230 Vac wall adapter power supply. Includes USA, UK, AUS, Euro adapter plugs and operation manual and plastic latching carry case, Display: 6 Digit Numeric /5 Digit Alphanumeric LCD, Tachometer Mode: 0-999,999 RPM
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT STROBOSCOPE LED 115/230VAC EURO PLUG 75840-624 6241-021 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty