Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proximity Sensors, Nidec Shimpo
Supplier: SEALS USA INC
Shimpo
SE-G2 DJ2-G MCS-3109 BI2-S12
75840-658EA 362.36 USD InStock
75840-658 75840-674 75840-696 75840-644
Proximity Sensors, Nidec Shimpo
Data Loggers
Ideal for gear speed measurement.
  • Rugged Proximity Sensors to measure position, movement, or rotational speeds
  • Economical solution for detecting metal in front of a sensing surface
  • Metal objects adjacent to or behind will not normally be detected
Model DJ2 features a pre-wired 17' cable assembly, zero-sensing capability, and small package size
Model B12-S12 is a three-wire DC normally open NPN (sinking) type proximity switch which features a short circuit and overload protection plus LED output indicator.
Model SE-G is a solid state proximity switch especially designed for gear sensing. This sensor will provide a square wave signal output and indicate detection via LED indicator on rear of unit. Maximum switching frequency is 8 kHz with a maximum sensing distance of 1.0mm. Gear pitch must be between 25.4 (min.) and 16.93 (max.).
Model MCS-3109 is a complimentary (NPN-NO and NPN-NC) proximity sensor normally used in high-vibration areas and where a greater sensing distance is required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty