Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Rotary Pulse Encoders, Nidec Shimpo

Supplier: SEALS USA INC
Shimpo
RE2B-600F RE1B-1000F RE1B-600F RE1B-60F RE1-BRKT RE2B-30F BRACKET-M18
75840-668EA 448.48 USD InStock
75840-668 75840-646 75840-648 75840-688 75840-670 75840-672 75840-650
Rotary Pulse Encoders, Nidec Shimpo
Data Loggers
Encoders are ideal for use with Panel Mount Tachometers and Counters to monitor motor operation.

  • Rotary Encoders available from 60 to 1000ppr, with 5000rpm maximum capability
  • Quadrature versions available
  • RE Series Encoders are ideal for usage with SHIMPO’s Panel Mount Tachometers and Counters

RE1B-60F: 60 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE1B-600F: 600 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE1B-1000F: 1000 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE2B-30F: Dual (90° out of phase) digital pulses for quadrature applications, 30 pulses/rev., maximum speed of 5000 rpm
RE2B-600F: Dual (90° out of phase) digital pulses for quadrature applications, 600 pulses/rev., maximum speed of 5000 rpm

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty