Would you like to log in again?
Contrast Sensor, Nidec Shimpo, SEALS USA INC
Supplier: SEALS USA INC
Shimpo
CNTR-1
75840-694EA 391.79 USD InStock
75840-694
Contrast Sensor, Nidec Shimpo, SEALS USA INC
Data Loggers
The CNTR effectively detects contrast differences between many colors on various surfaces over a range of 50 grayscale levels.
  • Fast response combined with small spot size allows for high-speed operation
  • Digital NPN/PNP output is automatically detected, plus analog output produces communication versatility
  • Learning function combined with dual color LED programming indication enables quick set up
  • Compact, IP65 enclosure permits usage in a variety of applications
  • Two years warranty
The CNTR sensor combines small spot size and fast response to achieve high-speed contrast detection capable of detecting a wide variety of marks and objects. The modulated white light source in the CNTR is focused to a 2.5 - 3 mm spot and directed toward a target. The diffused light returning to the sensor is measured. When the level is equal to or exceeds the threshold the discrete output changes state. Due to the fast response, it is suitable for use in high-speed applications. The sensor provides a discrete output that automatically configures to NPN and PNP. A PLC or a computer can be used to monitor the status of the discrete output signal.
Output: Auto-Detect PNP/NPN to 40 kHz + Analog 0-5 V
Sensing Distance: 1.1" (28 mm)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty