Would you like to log in again?
Cryokit 400, Cryogenic Gloves, Mapa Professional
Supplier: Mapa Professionnel
CRYKIT400410ZQK CRYKIT400407ZQK CRYKIT400411ZQK CRYKIT400408ZQK CRYKIT400409ZQK
75840-728EA 258.66 USD InStock
75840-728 75840-734 75840-736 75840-730 75840-732
Cryokit 400, Cryogenic Gloves, Mapa Professional
Gloves
These cryogenic waterproof glove are made of elastic and laminated blue fabric, internal polyester multilayer fleece and polyolefin porelle membrane offer dexterity in very low temperatures.
  • Interior finish with knitted thermal protection
  • Provide dexterity, comfort and also protection in cryogenic environments
  • Polyolefin Porelle membrane keeps hands dry while allowing air to flow
These gloves are suitable for applications handling liquid nitrogen and other cryogenic gases to protect from cold contact and prevent burns from liquid gas leakage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty