Would you like to log in again?
UHPLC Filters, Thermo Scientific
Supplier: Thermo Scientific
22180 22185 27006
75840-748PK 286.12 USD InStock
75840-748 75840-750
UHPLC Filters, Thermo Scientific
Filters Precolumn Filters and Frits
These UHPLC filters with low volume filter cartridge design, protect columns for Hypersil GOLD™ 1.9 µm and Syncronis 1.7 µm columns.
  • Maintain peak shape
  • Provide a low-volume filter cartridge design
  • Ensure minimal efficiency loss through dispersion
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty