Would you like to log in again?
All-In-One™ Containers for Gravity Compounding, Baxter IV Solutions
Supplier: Baxter IV Solutions
2B8134
75841-006CS 2566.36 USD InStock
75841-006
All-In-One™ Containers for Gravity Compounding, Baxter IV Solutions
Sample Containers General Purpose Containers
All-in-One™ container, empty ethylene vinyl acetate container with three lead transfer set for total nutrient admixtures.
  • Made of ethylene-vinyl acetate (EVA)
  • Sterile fluid path
  • Non-pyrogenic
Used in the compounding and storage of parenteral nutrition solutions prior to and during intravascular administration to a patient.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty