Would you like to log in again?
Streptavidin Blocker/Diluent, Enzo Life Sciences
Supplier: Enzo Life Sciences
ACC109-0100
75841-146EA 140 USD InStock
75841-146
Streptavidin Blocker/Diluent, Enzo Life Sciences
Buffers Electrophoresis Buffers Blocking Buffers
Ready-to-use blocking buffer and diluent, for use with SAVIEW® Plus detection reagents.
  • Prevent non-specific binding in biotin-conjugated detection systems
  • Intended for qualitative immunohistochemistry
  • May also be used with blood smears, cytosmears, and cell preparations
The Streptavidin Blocker/Diluent can be used to prevent non-specific binding in biotin-conjugated detection systems for IHC and ISH. It can also be used as a diluent for biotinylated antibodies. This product is intended for qualitative immunohistochemistry and in situ hybridization with formalin-fixed paraffin-embedded tissue sections and frozen sections. The blocker/diluent may also be used with blood smears, cytosmears, and cell preparations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty