Would you like to log in again?
Cryokit 550, 22" Cryogenic Gloves, Mapa Professional
Supplier: Mapa Professionnel
CRYKIT550558ZQK CRYKIT550551ZQK CRYKIT550559ZQK CRYKIT550550ZQK
75841-232EA 315.39 USD InStock
75841-232 75841-230 75841-236 75841-234
Cryokit 550, 22" Cryogenic Gloves, Mapa Professional
Gloves
These gloves are designed for applications handling liquid nitrogen and other cryogenic gases, to protect from cold contact and prevent burns from liquid gas leakage.
  • Multilayer fleece lining
  • Ventilation technology
  • Waterproof
Gloves remain ventilated due to the inner polyolefin porelle membrane which allows for air to flow and keeps hands dry.
A special elastic laminated blue fabric provides dexterity and comfort in the glove and cuff areas.
Certifications: CE Cat. III 0498 marked. Cold Hazards EN511: 231. Mechanical Hazards EN388: 2422.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty