Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Bel-Art H-B DURAC® Bi-Metallic Dial Thermometers, Bel-Art Products, a part of SP

Supplier: Bel-Art Products, a part of SP

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

B61310-2900 B61310-4600 B61310-4500 B61310-3500 B61310-5500 B61310-6100 B61310-3000 B61310-3300 B61310-4000 B61310-3600 B61310-6200 B61310-3200 B61310-4400 B61310-4800 B61310-6400 B61310-4100 B61310-6500 B61310-2800 B61310-3400 B61310-5700
75842-710CS 818.01 USD InStock
75842-710 75841-290 75841-294 75842-704 75841-292 75842-706 75841-298 75841-320 75842-718 75842-708 75852-862 30617-788 75841-302 75841-322 75841-300 75841-316 75841-306 75841-314 75841-308 89404-915
SP Bel-Art H-B DURAC® Bi-Metallic Dial Thermometers, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
Minimum immersion is 50mm (2”) in liquids or 100mm (4”) in gases.

  • External adjustment nut allows for simple field calibration

Thermometers feature polycarbonate lens and probe cover with pocket clip (except 44mm (1.7”) and 50mm (2”) dial sizes).

Each thermometer features accuracy to 1%, stainless steel case and stem and is individually packaged with a multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Test and Calibration.

The 44mm (1.7”) and 50mm (2”) dial sizes feature a beaker clip with built in adjustment tool in place of the probe cover with pocket clip.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty