Would you like to log in again?
Brew Plate™ Prepared Media Plates and Tubes, Bioplast Manufacturing
Supplier: Bioplast Manufacturing
BREW-BULK-ANBP BREW-BULK-YAQP BREW-BULK-YQPL BREW-BULK-HLPT BREW-BULK-YABP BREW-BULK-AEBP
75842-688PK 110.53 USD InStock
75842-688 75842-690 75842-692 75842-694 75842-684 75842-686
Brew Plate™ Prepared Media Plates and Tubes, Bioplast Manufacturing
Media for Microbiology Prepared Media for Microbiology
The collection of prefilled, easy-to-use beer testing bulk plates and tubes detect microbes that spoil beer during the brewing process, saving the brewer valuable time and money.
  • Allows the brewer to detect a broad collection of wild yeast strains
  • Plated and tubed agar
  • Send in the sample to Brew Plate Laboratories for microbe identification
Yeast Quad-Plate contains Lin's Cupric Sulfate Media, Lin's Wild Yeast Media, Lysine Media and Wallerstein Nutrient Media.
Yeast and Aerobic Quad-Plate contains Lin's Cupric Sulfate Media, Lee's Mulit-Differential Media, Lin's Wild Yeast Media and Wallerstein Nutrient Media.
Aerobic Bi-Plate contains Lee's Multi-Differential Media and MacConkey Agar.
Anaerobic Bi-Plate contains Lee's Multi-Differential Agar and Hsu's Lactobacillus/Pediococcus Media.
HLP Tubes are pre-filled with Hsu's Lactobacillus/Pediococcus Media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty