Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

The Brew Plate™ Digital Incubator 115 V, Bioplast Manufacturing

Supplier: Bioplast Manufacturing

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BREW-DIGINCU-H
75842-702EA 932.17 USD InStock
75842-702
The Brew Plate™ Digital Incubator 115 V, Bioplast Manufacturing
Incubators Standard Incubators
With digital temperature control, the Brew Plate™ Incubator provides convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob

  • Digital temperature control
  • Personal sized, 13 x 14.5" footprint (20L)
  • Internal outlet

Simply choose the desired temperature and the incubator immediately begins to heat up while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel.

The perfect, cost effective solution for any brewer.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty