Would you like to log in again?
Cell Stimulation and Protein Transport Inhibitor Cocktail, Biogems
Supplier: BioGems
63510-00-100
75843-426EA 116.38 USD InStock
75843-426
Cell Stimulation and Protein Transport Inhibitor Cocktail, Biogems
Assays Cellular Assays
The cell stimulation and protein transport inhibitor cocktail is a pre-mixed cocktail used to induce cytokine production and inhibit their intracellular transport for flow cytometry detection.
The solution is composed of an optimized mixture of brefeldin A, ionomycin, monensin, and PMA. PMA and ionomycin induce cytokine production in many cell types and brefeldin A and monensin are protein transport inhibitors.
The cell stimulation and protein transport inhibitor cocktail is supplied at a 500X concentration and should be used at 1X final concentration for cell stimulation. It can be added directly to the cell culture medium at 2 µl/ml. It has been tested by the stimulation of mouse splenocytes and subsequent intracellular staining and flow cytometry analysis of IFN-gamma.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty