Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD146 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1H12]

Supplier: BioGems
18512-60-100
75844-412EA 595.65 USD InStock
75844-412
Anti-CD146 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1H12]
Antibodies
The P1H12 monoclonal antibody reacts with CD146, 118kDA transmembrane glycoprotein that is also called the melanoma cell adhesion molecule (Mel-CAM), MUC18, Endo-CAM, and S-Endo. It is a prominently glycosylated protein with carbohydrates comprising the majority of its mass and it is expressed on melanoma, stromal, endothelial, and bone marrow fibroblasts. The P1H12 antibody cross reacts with human, mouse, canine, rabbit to the exclusion of rat protein.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: P1H12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty