Would you like to log in again?
pH Buffer Solutions, NIST/DIN, Mettler Toledo
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
51350054 30111139 30111136 51350058 30111138 51350056 30111137
11278-348EA 49 USD InStock
11278-348 75845-198 11278-354 11278-352 75845-224 75845-222 75845-200
pH Buffer Solutions, NIST/DIN, Mettler Toledo
Buffers pH Calibration Buffers
The NIST/DIN pH buffers are specified to three decimal places, for example 9.180, offering the highest possible pH precision.
  • Used for calibration/verification purposes
A detailed test certificate is available for every bottle and guarantees the tested values and traceability.
Certifications: NIST traceable.
Packaging: Packed in either sachets or bottles.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty