Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Petri Dishes, Glass
Supplier: VWR
248.202.03B 248.205.03A 248.207.03B 248.205.03B 248.202.04 248.202.03C 248.205.03C 248.207.04 248.202.03A 248.207.03C 248.205.00 248.207.03A 248.202.00
75845-522CS 168.24 USD InStock
75845-522 75845-544 75845-554 75845-520 75845-542 75845-552 75845-548 75845-526 75845-514 75845-546 75845-524 75845-512 75845-550
VWR® Petri Dishes, Glass
Culture Dishes Petri Dishes
These Petri dishes are suitable for use in pharmaceuticals, food processing, hospital environment and other industries for samples purposes.
  • Bottom and cover are flat inside and out, free from bubbles and streaks
  • Allow even distribution of culture media
  • Distortion-free transparency
  • Excellent temperature and chemical resistance
For quick identification, covers are inscribed in white with the word “cover” and bottoms are inscribed in blue with the word “bottom".
Petri dishes are made from high-quality type 1 class A, 33 expansion borosilicate glass.
Ordering information: Can be purchased as complete set (cover + bottom) or as replacement parts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty