Would you like to log in again?
BCYE Selective Agar (Buffered Charcoal Yeast Extract) with CCVC Antibiotics
Supplier: Hardy Diagnostics
G170
75845-728PK 121.92 USD InStock
75845-728
BCYE Selective Agar (Buffered Charcoal Yeast Extract) with CCVC Antibiotics
Media for Microbiology Dehydrated Media for Microbiology
Buffered Charcoal Yeast Extract (BCYE) selective agar with Cephalothin, Colistin, Vancomycin, and Cycloheximide antibiotics for the selective isolation and cultivation of Legionella from environmental and clinical specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty