Would you like to log in again?
ONPG, Rapid Test Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
R92
75845-730PK 106.14 USD InStock
75845-730
ONPG, Rapid Test Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the rapid determination of beta-Galactosidase activity of gram negative bacilli.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty