Would you like to log in again?
VWR® Flexible Multi-Tube Racks
Supplier: VWR
VWR®
HS120298 HS120297 HS120295 HS120300 HS120293 HS120294 HS120299 HS120296
75846-072CS 150.76 USD InStock
75846-072 75845-990 75846-000 75845-994 75845-992 75845-988 75845-998 75845-996
VWR® Flexible Multi-Tube Racks
Racks Tube Racks
This rack is truly adaptable, as each well adjusts to accommodate 7 to 50 mL tube sizes and holds them in a firm upright position.
  • Racks can be linked together for additional capacity
  • Open sides allow for label viewing
  • Flat base to hold tubes upright
  • Accommodates tubes from 7 to 50 mL sizes
  • Autoclavable
VWR® Flexible Multi-Tube Racks feature unique slots that adapt to accommodate varied tubes sizes and hold them in a firm upright position. Racks are manufactured from POM, and can be linked together for additional holding capacity. Linked racks should always be supported underneath when carried or transported. They are available in solid colors, or color combinations to simplify coding identification. Each individual rack holds four tubes. Supplied two racks per pack.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty