Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Roc C18 HPLC Columns (USP L1), Restek

Supplier: Restek
9534515 9534365 953436E 9534565 9534315 953457E 953431E 953456E 9534575 953451E
75846-150EA 424.74 USD InStock
75846-150 75846-152 75846-140 75846-154 75846-142 75846-156 75846-144 75846-158 75846-146 75846-138
Roc C18 HPLC Columns (USP L1), Restek
Chromatography Columns
The general-purpose Restek C18 is a conventional monomeric octadecylsilane column suitable for analyses of a wide range of compounds from acidic through slightly basic.

  • USP phase code: L1
  • LigandtType: monomeric C18
  • Surface area: 300 m²/g
  • Carbon load: 20%
  • End-cap: yes
  • pH range: 2.5 to 8.0
  • Maximum temperature: 80 °C

Roc HPLC columns are built to be the cornerstone for your LC lab. These solid, reliable columns are pressure rated for any 400 bar HPLC system and deliver the peak shape, reproducibility, ruggedness, and performance you demand for all of your conventional HPLC applications. An exceptional value, Roc columns are priced for bulk use in routine analyses and large-volume workflows.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty