Would you like to log in again?
AquaSampler, Bürkle
Supplier: Bürkle
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5308-1050 5308-0700 5308-0350
75846-218EA 763.24 USD InStock
75846-218 75846-240 75846-238
AquaSampler, Bürkle
Samplers Sample Dippers
Sampler for ground water samples from wells, pits, and other sources
  • FEP/PTFE ultra pure, anti-adhesive surface
  • Supplied without cable
Fits all normal well shafts with control openings to a diameter of 2", without tilting or getting stuck. Ultra pure materials guarantee the highest degree of sample purity. Easy to clean thanks to rounded edges and threads.
Packaging: Supplied one sampler per package.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty