Would you like to log in again?
Dipping Bottle, Bürkle
Supplier: Bürkle
5365-4000
75846-222EA 1112.27 USD InStock
75846-222
Dipping Bottle, Bürkle
Bottles Water Sampling Bottles
Dipping bottle with glass container for sampling in sewage plants, as well as mud and water samples.
  • Equipped with Easy-Click; a safe and practical closure for a rapid bottle exchange
  • High weight (3.3 kg/7.28 lbs.) for fast lowering
  • Height with handle is 330 mm (12.99")
  • Tested for use in the Ex Protection Zone 0
For testing crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum, and liquid lubricants of Hazard Categories IIA and IIB. Constructed from completely non-sparking materials. Dipping frame made of nickel-plated brass, with an easy-click closure made of electrically conductive polyamide.
Packaging: Supplied one dipping bottle per package.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty