Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dipping vessel, Burkle

Supplier: Bürkle

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

5365-1000 5365-2000
75846-296EA 2430.54 USD InStock
75846-296 75846-228
Dipping vessel, Burkle
Samplers Fluid Samplers
Dipping vessel for taking all-layer samples from tanks, tank cars, water courses, water purification plants.

  • Equipped with Easy-Flow; a flow optimized flow valve
  • High weight (2.1 kg/4.63 lbs.) for faster lowering
  • Height with handle is 427 mm (16.81")
  • Made of non-sparking brass or stainless steel AISI 304 (1.4301) for Ex area
  • Tested for use in the Ex Protection Zone 0
  • Explosion Categories IIA, IIB, and IIC

Packaging: Supplied one dipping vessel per package.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty