Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HardyDisks™ AST Cefaclor, CEC-30, Hardy Diagnostics
Supplier: Hardy Diagnostics
HardyDisks™
Z8121
75846-316PK 21.89 USD InStock
75846-316
HardyDisks™ AST Cefaclor, CEC-30, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
Cefaclor, CEC-30, HardyDisks™ AST, for susceptibility testing.
  • Impregnated paper disks used for Antimicrobial Susceptibility Testing (AST)
  • Spring-loaded for smooth and reliable dispensing
  • Compatible with single or multi-disk dispensers
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.The last disk in each cartridge is clearly marked so as to alert you when refills are needed.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers as listed below.
Delivery information: Freeze on arrival.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty