Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Standard Muffle Furnaces, FO Series, Yamato

Supplier: Yamato

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FO-300CR FO-210CR FO-100CR FO-410CR FO-110CR FO-200CR FO-610CR FO-710CR FO-510CR FO-310CR FO-810CR
75846-648EA 7202.24 USD InStock
75846-648 75846-380 75846-382 75846-374 75846-384 75846-376 75846-386 75846-650 75846-378 75846-644 75846-646
Standard Muffle Furnaces, FO Series, Yamato
Ovens Furnaces (Ashing and Muffle Ovens)
The Standard Muffle Furnace series features a wide selection of space-saving, compact units with maximum inner capacity.

  • Easy-to-Use Controller
  • Communication Port and Exhaust Port
  • Cooling Axial Fan Motor
  • High Temperature Accuracy at ±2.0 °C
  • Operating Temperature Range: 100 to ~1150 °C

Cold-rolled steel plate exterior with a baked-on melamine resin finish and ceramic fiber interior.

A digital display features UP/DOWN keys for setting the temperature. Functions include calibration offset and power failure compensation. Safety features include an electric leakage breaker and an overheat prevention device. Self-diagnostic functions ensure safe operation.

A firmly sealed chamber door for the iron-chrome wire heater results in excellent heat tightness. A lock and exhaust cap are included.

Programs: Maximum 6 Patterns: 30 Steps x 1 Pattern, 15 Steps x 2 Patterns, or 10 Steps x 3 Patterns

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty