Would you like to log in again?
Digital Laboratory Stirrer, Yamato
Supplier: Yamato
LR-500AA LR-500BA
75846-392KT 2848.69 USD InStock
75846-392 75846-394
Digital Laboratory Stirrer, Yamato
Stirrers Overhead Stirrers
This digital laboratory stirrer achieves high torque enabling stirring of high viscosity solution.
  • LED Overload Display with Two Steps
  • Speed Feedback Control
  • Shaft: 10 mm dia.
  • DC Brushless Motor Eliminates Brush Replacement
  • Maintenance-Free Operation
The direct-drive system reduces noise and requires little maintenance
An overload lamp turns on when the unit is exceeding the maximum load, stopping the motor automatically. Revolution feedback control function can maintain the setting rate despite change of load, which is especially suitable for high-viscosity samples.
The featured DC brushless motor has no brushes to cause sparks. The unit also features a digital tachometer for easy speed setting and confirmation. Load on the stirring shaft can be monitored by the LED display. To ensure safe operation, the unit stops when overloaded.
Ordering information: All models come as a set which includes main unit, stand, rod, stainless steel shaft, and 75 mm 4-blade propeller.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty